MilliporeSigma
  • Antiviral activity of palmitic acid via autophagic flux inhibition in zebrafish (Danio rerio).
Antiviral activity of palmitic acid via autophagic flux inhibition in zebrafish (Danio rerio).
Fish & shellfish immunology (2019-11-05)
Marta Librán-Pérez, Patricia Pereiro, Antonio Figueras, Beatriz Novoa
ABSTRACT
Fatty acids (FAs) are key elements that affect not only growth but also different immune functions, and therefore, nutrition is important for growing healthy fish. Zebrafish (Danio rerio) is a good model for assessing the beneficial effects of immunostimulants, including FAs, before applying them in aquaculture. Accordingly, this study evaluated the effects of palmitic acid (PA) treatment on different immune parameters of zebrafish and on the mortality caused by the spring viremia of carp virus (SVCV). The results suggest that PA modulates the infection outcome in vivo, which benefits zebrafish and results in reduced mortality and viral titres. The antiviral protection elicited by this FA seems to be associated with the inhibition of autophagy and is independent of other immune processes, such as neutrophil proliferation or type I interferon (IFN) activity. The use of PA as an immunostimulant at low concentrations showed great potential in the prevention of SVCV infections; therefore, this FA could help to prevent the mortality and morbidity caused by viral agents in aquacultured fish. Nevertheless, the potentially detrimental effects of suppressing autophagy in the organism should be taken into account.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
Anti-Rabbit IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
Palmitic acid, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685
Sigma-Aldrich
Triton X-100, laboratory grade
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
Please contact Customer Service for Availability
₩81,200
1 kg
Please contact Customer Service for Availability
₩107,387
10 kg
Please contact Customer Service for Availability
₩499,685