MilliporeSigma
  • Early inflammatory reactions in atherosclerosis are induced by proline-rich tyrosine kinase/reactive oxygen species-mediated release of tumor necrosis factor-alpha and subsequent activation of the p21Cip1/Ets-1/p300 system.
Early inflammatory reactions in atherosclerosis are induced by proline-rich tyrosine kinase/reactive oxygen species-mediated release of tumor necrosis factor-alpha and subsequent activation of the p21Cip1/Ets-1/p300 system.
Arteriosclerosis, thrombosis, and vascular biology (2011-03-05)
Asako Katsume, Mitsuhiko Okigaki, Akihiro Matsui, Jishan Che, Yasushi Adachi, Eigo Kishita, Shinichiro Yamaguchi, Koji Ikeda, Tomomi Ueyama, Satoaki Matoba, Hiroyuki Yamada, Hiroaki Matsubara
ABSTRACT
Reactive oxygen species (ROS) are involved in the initial process of atherosclerosis, whereas it remains to be determined how atherogenic stimulus causes ROS-mediated proinflammatory reactions. Here, we focused on proline-rich tyrosine kinase (PYK2)-mediated ROS generation and examined how atherogenic stimulus causes early proinflammatory reactions. PYK2-deficient (knockout [KO]) (PYK2-KO) mice were crossbred with apolipoprotein E (ApoE)-deficient (PYK2-KO/ApoE-KO) mice. PYK2-KO/ApoE-KO mice and endothelial cells (EC) were used for the study. Aortic atherogenic lesions in PYK2-KO/ApoE-KO mice were markedly decreased (55% versus ApoE-KO) after 8 weeks of a Western diet. Aortic PYK2 was activated as early as 7 days after the Western diet, when inflammatory cells were not yet activated. Addition of the proatherogenic oxidized phospholipid lysophosphatidylcholine caused activation of endothelial PYK2. Lysophosphatidylcholine-activated PYK2 induced NADPH oxidase-mediated ROS generation and ROS-mediated synthesis of tumor necrosis factor-α (TNFα), vascular cell adhesion molecule-1 (VCAM-1), monocyte chemotactic protein-1 (MCP-1), and p21Cip1/Ets-1. Neutralizing anti-TNFα antibody or knockdown of p21Cip1/Ets-1 system blocked the induction of VCAM-1 and MCP-1. PYK2 deficiency abolished these ROS-mediated proinflammatory reactions. Further analysis revealed that PYK2/ROS-mediated p21Cip1/Ets-1 activation upregulated the transcription of the MCP-1 gene in collaboration with p300 transcription coactivator. PYK2 is a key tyrosine kinase activated by high cholesterol exposure, which causes ROS-mediated TNFα release and induces TNFα-dependent expression of proinflammatory molecules via the p21Cip1/Ets-1/p300 transcription system.
Sigma-Aldrich
1-Stearoyl-sn-glycero-3-phosphocholine, ≥99%, powder