Merck
  • Liposome-entrapped GABA modulates the expression of nNOS in NG108-15 cells.
Liposome-entrapped GABA modulates the expression of nNOS in NG108-15 cells.
Journal of neuroscience methods (2016-10-25)
Gisele C Vaz, Neeru M Sharma, Hong Zheng, Matthew C Zimmerman, Robson S Santos, Frederic Frezard, Marco A P Fontes, Kaushik P Patel
ABSTRACT
Liposomes are concentric lipid vesicles that allow a sustained release of entrapped substances. GABA (γ-aminobutyric acid) is the most prevalent inhibitory neurotransmitter in the central nervous system. Using GABA-containing liposomes (GL) prepared by the freeze-thawing method, we determined the effect of sustained release of GABA on expression of neuronal nitric oxide synthase (nNOS) and GABAA receptor (GABAAR) in an in vitro neuronal model. Neuronal cell line NG108-15 treated with different doses of GL during 24h showed an increase in expression of GABAAR (54 and 50% with 10 and 20ng doses, respectively) and nNOS (138, 157 and 165% with 20, 50 and 100ng doses, respectively) compared with cells treated with empty liposomes (EL). Additionally, cells treated with 50ng of GL showed an increase in GABAAR (23%) after 1h followed by an increase in nNOS (55, 46 and 55%) at 8, 12 and 24h time points, respectively. Immunofluorescence experiments confirmed an increase in nNOS (134%) and basal intracellular levels of nitric oxide (84%) after GL treatment. Further, treatment of cells with GL showed a decrease in expression of a protein inhibitor of nNOS (PIN) (26, 66 and 57% with 20, 50 and 100ng doses respectively) compared with control. This is first demonstration for the development of GL that allows sustained slow release of this neurotransmitter. These results suggest that a slow release of GABA can change the expression of nNOS possibly via alteration in PIN levels in neuronal cells.
Sigma-Aldrich
Anti-GABA(A) receptor subunit gamma-2 Antibody, serum, from rabbit