MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • A novel UMOD gene mutation associated with uromodulin-associated kidney disease in a young woman with moderate kidney dysfunction.
A novel UMOD gene mutation associated with uromodulin-associated kidney disease in a young woman with moderate kidney dysfunction.
Internal medicine (Tokyo, Japan) (2015-03-20)
Akihiro Kuma, Masahito Tamura, Nana Ishimatsu, Tetsu Miyamoto, Ryota Serino, Shingo Ishimori, Naoya Morisada, Kazumoto Iijima, Sohsuke Yamada, Masaaki Takeuchi, Haruhiko Abe, Yutaka Otsuji
ABSTRACT
Uromodulin-associated kidney disease (UAKD) is an autosomal dominant disease caused by a mutation in the uromodulin (UMOD) gene, leading to end-stage renal disease. We herein report the case of a family with UAKD caused by a novel mutation (C135G) in the UMOD gene. A 31-year-old woman had a low estimated glomerular filtration rate (59.7 mL/min per 1.73 m(2)). Her father, grandfather and paternal aunt had received maintenance hemodialysis therapy since their 40's. This case underscores the importance of performing genetic testing in young patients even in cases involving only moderate abnormalities in the kidney function.
Sigma-Aldrich
Anti-UMOD antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution