Merck
  • Alpha interferon and ribavirin combination therapy of chronic hepatitis D.
Alpha interferon and ribavirin combination therapy of chronic hepatitis D.
Antimicrobial agents and chemotherapy (2005-02-25)
Sabahattin Kaymakoglu, Cetin Karaca, Kadir Demir, Sule Poturoglu, Ahmet Danalioglu, Selim Badur, Murvet Bozaci, Fatih Besisik, Yilmaz Cakaloglu, Atilla Okten
ABSTRACT
The success of alpha interferon (IFN-alpha) monotherapy for the treatment of chronic hepatitis D is very limited. In this study, the efficacy of IFN-alpha and ribavirin combination therapy for chronic hepatitis D was investigated. Nineteen patients (15 males; mean age +/- standard deviation, 36.8 +/- 12.8 years) with chronic hepatitis D who were treated with IFN-alpha2b (10 million U, three times/week, subcutaneously) and ribavirin (1,000 to 1,200 mg/day, orally) for 24 months were studied. All patients had compensated liver disease (15 were precirrhotic), elevated transaminase levels, and hepatitis D virus RNA positivity at baseline. Genotypic analyses revealed hepatitis D virus genotype I and hepatitis B virus genotype D. All patients completed the 24 months of treatment and at least 6 months (7 to 19 months) of a follow-up period. Biochemical responses were observed in eight patients (42.1%) at the end of treatment and in seven patients (36.8%) at the end of follow-up. Eight patients (42.1%) at the end of treatment and four patients (21%) at the end of follow-up had virological responses. In conclusion, combination treatment of IFN-alpha and ribavirin for chronic hepatitis D is not able to induce virological responses at a sufficient rate, despite its partial effectiveness in improving biochemical responses, and is not superior to IFN-alpha monotherapy.
Sigma-Aldrich
Ribonuclease inhibitor human, Inhibits ribonucleases during transcription and translation experiments