MilliporeSigma
  • Celecoxib restores angiogenic factor expression at the maternal-fetal interface in the BPH/5 mouse model of preeclampsia.
Celecoxib restores angiogenic factor expression at the maternal-fetal interface in the BPH/5 mouse model of preeclampsia.
Physiological genomics (2018-03-10)
Dorien Reijnders, Chin-Chi Liu, Xinjing Xu, Anna M Zhao, Kelsey N Olson, Scott D Butler, Nataki C Douglas, Jenny L Sones
ABSTRACT
Preeclampsia (PE), a hypertensive disease of pregnancy, is a leading cause of fetal and maternal morbidity/mortality. Early angiogenic and inflammatory disturbances within the placenta are thought to underlie the development of the maternal PE syndrome and poor pregnancy outcomes. However, the exact etiology remains largely unknown. Here, we use the BPH/5 mouse model of PE to elucidate the way in which inflammation early in pregnancy contributes to abnormal expression of angiogenic factors at the maternal-fetal interface. We have previously described improvement in maternal hypertension and fetal growth restriction in this model after treatment with the anti-inflammatory cyclooxygenase-2 (Cox2) specific inhibitor celecoxib. To further characterize the mechanisms by which celecoxib improves poor pregnancy outcomes in BPH/5 mice, we determined expression of angiogenic factors and complement pathway components after celecoxib. In BPH/5 implantation sites there was increased hypoxia inducible factor-1α ( Hif1α), heme oxygenase-1 ( Ho-1), and stem cell factor ( Scf) mRNA concomitant with elevated prostaglandin synthase 2 ( Ptgs2), encoding Cox2, and elevated VEGF protein. Angiopoietin 1 ( Ang1), tunica interna endothelial cell kinase-2 receptor ( Tie2), complement factor 3 ( C3), and complement factor B ( CfB) were increased in midgestation BPH/5 placentae. Whereas BPH/5 expression levels of VEGF, Ang1, and Tie2 normalized after celecoxib, placental C3 and CfB mRNA remained unchanged. However, celecoxib did reduce the pregnancy-specific circulating soluble fms-like tyrosine kinase-1 (sFlt-1) rise in BPH/5 mice at midgestation. These data show that elevated Cox2 during implantation contributes to placental angiogenic factor imbalances in the BPH/5 mouse model of PE.
Sigma-Aldrich
Anti-Vascular Endothelial Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder