MilliporeSigma
In vitro human nail penetration and kinetics of panthenol.
International journal of cosmetic science (2008-05-21)
X Hui, S B Hornby, R C Wester, S Barbadillo, Y Appa, H Maibach
ABSTRACT
The in vitro absorption of panthenol into and through the human nail was examined in this study. Panthenol, the alcohol form of pantothenic acid (vitamin B5), is believed to act as a humectant and improve the flexibility and strength of nails. A liquid nail treatment formulated with panthenol (2%) was compared to a solution of panthenol (2%) in water. Fingernail specimens were dosed daily for 7 days with either the nail treatment (non-lacquer film forming) formulation or aqueous solution with sampling performed every 24 h. Panthenol concentrations were determined in the dorsal surface, interior (by drilling and removal) and in the supporting bed under the human nail. Panthenol levels in the dorsal nail (R(2) = 0.87; P < 0.001), nail interior (R(2) = 0.94; P < 0.001) and nail supporting bed (R(2) = 0.79; P < 0.003) showed a significant linear increase with each day of dosing. Significantly more panthenol was delivered into the interior nail and supporting bed by a nail treatment formulation than from an aqueous solution. The film acts not only as a reservoir of panthenol, but also acts to increase the hydration of the nail and the thermodynamic activity of panthenol as well, thereby enhancing diffusion.
Sigma-Aldrich
D-Pantothenic acid hemicalcium salt, ≥98.0%
Sigma-Aldrich
D-Panthenol, ≥98.0% (NT)