MilliporeSigma
EGFR/MDM2 signaling promotes NF-κB activation via PPARγ degradation.
Carcinogenesis (2016-01-01)
Ying Xu, Jianhua Jin, Wenbo Zhang, Zhi Zhang, Jiaming Gao, Qian Liu, Chenglin Zhou, Qinggang Xu, Haifeng Shi, Yongzhong Hou, Juanjuan Shi
ABSTRACT
Dysregulated expression of epidermal growth factor receptor (EGFR) has been implicated in many cancer events, while peroxisome proliferator-activated receptor γ (PPARγ) negatively regulates cancer progression. The molecular mechanism of EGFR interaction with PPARγ is still unclear. Here, we found that nuclear EGFR induced phosphorylation of PPARγ at Tyr-74 leading to PPARγ ubiquitination and degradation by mouse double minute 2 (MDM2) ubiquitin ligase. PPARγ degradation by EGFR/MDM2 signaling resulted in accumulation of nuclear factor-kappaB (NF-κB)/p65 protein levels and increasing NF-κB activation. In contrast, PPARγ-Y74A mutant reversed this event. Moreover, PPARγ-Y74A mutant suppressed cell proliferation and increased chemotherapeutic agent-induced cancer cell sensitivity. Importantly, the clinical findings show that the nuclear phosphorylation of PPARγ-Y74 and EGFR expression in colonic cancer tissues was higher than that in control normal tissues. Thus, our study revealed a novel molecular mechanism that nuclear EGFR/NF-κB signaling promoted cell proliferation by destructing PPARγ function, which provides a novel strategy for cancer treatment.
Sigma-Aldrich
Anti-Phosphotyrosine antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution