MilliporeSigma
  • Identification of proteins cleaved downstream of caspase activation in monocytes undergoing macrophage differentiation.
Identification of proteins cleaved downstream of caspase activation in monocytes undergoing macrophage differentiation.
The Journal of biological chemistry (2006-04-26)
Séverine Cathelin, Cédric Rébé, Lamya Haddaoui, Nicolas Simioni, Frédérique Verdier, Michaëla Fontenay, Sophie Launay, Patrick Mayeux, Eric Solary
ABSTRACT
We have shown previously that caspases were specifically involved in the differentiation of peripheral blood monocytes into macrophages while not required for monocyte differentiation into dendritic cells. To identify caspase targets in monocytes undergoing macrophagic differentiation, we used the human monocytic leukemic cell line U937, whose macrophagic differentiation induced by exposure to 12-O-tetradecanoylphorbol 13-acetate (TPA) can be prevented by expression of the baculovirus caspase-inhibitory protein p35. A comparative two-dimensional gel proteomic analysis of empty vector- and p35-transfected cells after 12 h of exposure to 20 nm TPA, followed by mass spectrometry analysis, identified 38 differentially expressed proteins. Those overexpressed in p35-expressing cells (n = 16) were all full-length, whereas half of those overexpressed in control cells (n = 22) were N- or C-terminal cleavage fragments. The cleavage or degradation of seven of these proteins was confirmed in peripheral blood monocytes undergoing macrophage colony-stimulating factor-induced macrophagic differentiation. In U937 cells exposed to TPA, these proteolytic events can be inhibited by expression of a caspase-8 dominant negative mutant or the cowpox virus CrmA caspase inhibitor. These cleavages provide new insights to analyze the role of caspases in this specific differentiation program.
Sigma-Aldrich
Monoclonal Anti-hnRNP-C1/C2 antibody produced in mouse, clone 4F4, purified immunoglobulin, buffered aqueous solution