MilliporeSigma
  • A Multiplexed System for Quantitative Comparisons of Chromatin Landscapes.
A Multiplexed System for Quantitative Comparisons of Chromatin Landscapes.
Molecular cell (2015-12-22)
Peter van Galen, Aaron D Viny, Oren Ram, Russell J H Ryan, Matthew J Cotton, Laura Donohue, Cem Sievers, Yotam Drier, Brian B Liau, Shawn M Gillespie, Kaitlin M Carroll, Michael B Cross, Ross L Levine, Bradley E Bernstein
ABSTRACT
Genome-wide profiling of histone modifications can provide systematic insight into the regulatory elements and programs engaged in a given cell type. However, conventional chromatin immunoprecipitation and sequencing (ChIP-seq) does not capture quantitative information on histone modification levels, requires large amounts of starting material, and involves tedious processing of each individual sample. Here, we address these limitations with a technology that leverages DNA barcoding to profile chromatin quantitatively and in multiplexed format. We concurrently map relative levels of multiple histone modifications across multiple samples, each comprising as few as a thousand cells. We demonstrate the technology by monitoring dynamic changes following inhibition of p300, EZH2, or KDM5, by linking altered epigenetic landscapes to chromatin regulator mutations, and by mapping active and repressive marks in purified human hematopoietic stem cells. Hence, this technology enables quantitative studies of chromatin state dynamics across rare cell types, genotypes, environmental conditions, and drug treatments.
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys4) Antibody, Upstate®, from rabbit
Sigma-Aldrich
KDM5-C70, ≥95% (HPLC)
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit