Merck
  • Hippo Cascade Controls Lineage Commitment of Liver Tumors in Mice and Humans.
Hippo Cascade Controls Lineage Commitment of Liver Tumors in Mice and Humans.
The American journal of pathology (2018-01-30)
Shanshan Zhang, Jingxiao Wang, Haichuan Wang, Lingling Fan, Biao Fan, Billy Zeng, Junyan Tao, Xiaolei Li, Li Che, Antonio Cigliano, Silvia Ribback, Frank Dombrowski, Bin Chen, Wenming Cong, Lixin Wei, Diego F Calvisi, Xin Chen
ABSTRACT
Primary liver cancer consists mainly of hepatocellular carcinoma (HCC) and intrahepatic cholangiocarcinoma (ICC). A subset of human HCCs expresses a ICC-like gene signature and is classified as ICC-like HCC. The Hippo pathway is a critical regulator of normal and malignant liver development. However, the precise function(s) of the Hippo cascade along liver carcinogenesis remain to be fully delineated. The role of the Hippo pathway in a murine mixed HCC/ICC model induced by activated forms of AKT and Ras oncogenes (AKT/Ras) was investigated. The authors demonstrated the inactivation of Hippo in AKT/Ras liver tumors leading to nuclear localization of Yap and TAZ. Coexpression of AKT/Ras with Lats2, which activates Hippo, or the dominant negative form of TEAD2 (dnTEAD2), which blocks Yap/TAZ activity, resulted in delayed hepatocarcinogenesis and elimination of ICC-like lesions in the liver. Mechanistically, Notch2 expression was found to be down-regulated by the Hippo pathway in liver tumors. Overexpression of Lats2 or dnTEAD2 in human HCC cell lines inhibited their growth and led to the decreased expression of ICC-like markers, as well as Notch2 expression. Altogether, this study supports the key role of the Hippo cascade in regulating the differentiation status of liver tumors.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Protein Tyrosine Phosphatase µ Antibody, clone SBK15, ascites fluid, clone SBK15, Chemicon®