MilliporeSigma
  • Development of CAPER peptides for the treatment of triple negative breast cancer.
Development of CAPER peptides for the treatment of triple negative breast cancer.
Cell cycle (Georgetown, Tex.) (2020-01-15)
Shannon D Chilewski, Devyani Bhosale, Sundee Dees, Isaac Hutchinson, Rachel Trimble, Laura Pontiggia, Isabelle Mercier, Jean-Francois Jasmin
ABSTRACT
Triple negative breast cancer (TNBC) is a heterogeneous disease, which lacks expression of the estrogen receptor (ER), progesterone receptor (PR) and the human epidermal growth factor 2 receptor (HER2). This subtype of breast cancer has the poorest prognosis with limited therapies currently available, and hence additional options are needed. CAPER is a coactivator of the activator protein-1 (AP-1) (interacting specifically with the c-Jun component) and the ER and is known to be involved in human breast cancer pathogenesis. Recent published data have demonstrated a role for CAPER in TNBC and, as such, disrupting the function of CAPER with c-Jun could be a novel approach to treat TNBC patients. The data presented here shows the development and in vitro testing of CAPER-derived peptides that inhibit the coactivator activity of CAPER with c-Jun. These CAPER peptides result in a decrease in cell number and an increase in apoptosis in two TNBC cell lines, MDA-MB-231 and BT-549, while having no effect on the non-tumorigenic cell line MCF 10A. Additionally, two modes of action were demonstrated which appear to be cell line dependent: 1) a modulation of phosphorylated c-Jun leading to a decrease in Bcl-2 in MDA-MB-231 cells and a decrease in p21 in BT-549 cells and 2) a decrease in DNA repair proteins, leading to impaired DNA repair function in MDA-MB-231 cells. The data presented here supports further development of CAPER-derived peptides for the treatment of TNBC.
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 mg
Estimated to ship on March 23, 2025
$116.00
Sigma-Aldrich
Insulin solution from bovine pancreas, 10 mg/mL insulin in 25  mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 mg
Estimated to ship on March 23, 2025
$116.00
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 mg
Estimated to ship on March 23, 2025
$116.00
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 mg
Estimated to ship on March 23, 2025
$116.00