MilliporeSigma
  • Potassium bromate: Effects on bread components, health, environment and method of analysis: A review.
Potassium bromate: Effects on bread components, health, environment and method of analysis: A review.
Food chemistry (2019-12-23)
Venu Shanmugavel, Kotturu Komala Santhi, Anjali H Kurup, Sureshkumar Kalakandan, Arunkumar Anandharaj, Ashish Rawson
ABSTRACT
Potassium bromate, is an oxidizing agent and one of the best and cheapest dough improvers in the baking industry. Due to its positive effects it plays a major role in the bread-making industry. Potassium bromate has significant effect on food biomolecules, such as starch and protein, as it affects the extent of gelatinization, viscosity, swelling characteristics as well as gluten proteins; it removes the sulfhydryl group and leads to the formation of disulfide linkages and thus improves the bread properties. However, there are many reports elucidating its negative impact on human health. It is deemed as a potential human carcinogen by IARC and classified under class 2B. Due to this, countries across world have either partially or completely banned it. Numerous techniques have evolved to determine the concentration of potassium bromate in bread. This review explains in detail, the effects of potassium bromate on biomolecules, human health, environment and various methods of analysis.
Sigma-Aldrich
Calcium peroxide, 75%, −200 mesh
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 g
Estimated to ship on January 27, 2025
Details...
₩40,499
500 g
Estimated to ship on January 27, 2025
Details...
₩106,880
1 kg
Estimated to ship on January 27, 2025
Details...
₩121,597
50 kg
Estimated to ship on February 07, 2025
Details...
₩1,568,683