Merck
  • Chronic Light Exposure in the Middle of the Night Disturbs the Circadian System and Emotional Regulation.
Chronic Light Exposure in the Middle of the Night Disturbs the Circadian System and Emotional Regulation.
Journal of biological rhythms (2016-04-15)
Tomoko Ikeno, Lily Yan
ABSTRACT
In mammals, the circadian system is composed of a principal circadian oscillator located in the suprachiasmatic nucleus (SCN) and a number of subordinate oscillators in extra-SCN brain regions and peripheral tissues/organs. However, how the time-keeping functions of this multiple oscillator circuit are affected by aberrant lighting environments remains largely unknown. In the present study, we investigated the effects of chronic light exposure in the middle of the night on the circadian system by comparing the mice housed in a 12:4:4:4-h L:DLD condition with the controls in 12:12-h L:D condition. Daily rhythms in locomotor activity were analyzed and the expression patterns of protein products of clock genes Period1 and Period2 (PER1 and PER2) were examined in the SCN and extra-SCN brain regions, including the dorsal striatum, hippocampus, paraventricular nucleus (PVN), and basolateral amygdala (BLA). Following 2 weeks of housing in the L:DLD condition, animals showed disturbed daily rhythms in locomotor activity and lacked daily rhythms of PER1 and PER2 in the SCN. In the extra-SCN brain regions, the PER1 and PER2 rhythms were affected in a region-specific pattern, such that they were relatively undisturbed in the striatum and hippocampus, phase-shifted in the BLA, and abolished in the PVN. In addition, mice in the L:DLD condition showed increased anxiety-like behaviors and reduced brain-derived neurotropic factor messenger RNA expression in the hippocampus, amygdala, and medial prefrontal cortex, which are brain regions that are involved in emotional regulation. These results indicate that nighttime light exposure leads to circadian disturbances not only by abolishing the circadian rhythms in the SCN but also by inducing misalignment among brain oscillators and negatively affects emotional processing. These observations serve to identify risks associated with decisions regarding lifestyle in our modern society.
Sigma-Aldrich
Anti-PER2 Antibody, serum, Chemicon®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-mPER1 (residues 6-21) Antibody, serum, from rabbit