MilliporeSigma
  • Regulation of human erythrocyte Na+/H+ exchange by soluble and particulate guanylate cyclase.
Regulation of human erythrocyte Na+/H+ exchange by soluble and particulate guanylate cyclase.
The American journal of physiology (1996-11-01)
V Petrov, P Lijnen
ABSTRACT
Guanylate cyclase activity in human erythrocytes is investigated by evaluating the intracellular guanosine 3',5'-cyclic monophosphate (cGMP) content in the presence of various agents that exert specific effects on soluble or particulate guanylate cyclase. The increase in the intraerythrocyte cGMP content by the soluble guanylate cyclase activators nitroprusside and NaNO2 suggests the presence of this enzyme in human erythrocytes. The effects of four different atrial natriuretic peptide (ANP) fragments on the intraerythrocyte cGMP content is also studied. ANP II and ANP III increase the intraerythrocyte cGMP content, whereas ANP I and des-Ser5,des-Ser6-ANP III are ineffective. Thus our data show that human erythrocytes possess particulate guanylate cyclase together with the soluble enzyme. The ANP fragments ANP II and ANP III also activate the erythrocyte Na+/H+ exchange. Nitroprusside, M & B 22948 (an inhibitor of cGMP phosphodiesterase), and the cGMP analogues dibutyryl cGMP and 8-bromoguanosine 3',5'-cyclic monophosphate also increase the erythrocyte Na+/H+ exchange rate. The latter data also suggest that the erythrocyte Na+/H+ exchange is regulated by cGMP.
Sigma-Aldrich
Atriopeptin III rat / ANP 127-150 rat
Sigma-Aldrich
ANP 126-150 rat /Auriculin B, ANP 4-28 rat