Merck
  • Highly Combinatorial Genetic Interaction Analysis Reveals a Multi-Drug Transporter Influence Network.
Highly Combinatorial Genetic Interaction Analysis Reveals a Multi-Drug Transporter Influence Network.
Cell systems (2019-11-02)
Albi Celaj, Marinella Gebbia, Louai Musa, Atina G Cote, Jamie Snider, Victoria Wong, Minjeong Ko, Tiffany Fong, Paul Bansal, Joseph C Mellor, Gireesh Seesankar, Maria Nguyen, Shijie Zhou, Liangxi Wang, Nishka Kishore, Igor Stagljar, Yo Suzuki, Nozomu Yachie, Frederick P Roth
ABSTRACT
Many traits are complex, depending non-additively on variant combinations. Even in model systems, such as the yeast S. cerevisiae, carrying out the high-order variant-combination testing needed to dissect complex traits remains a daunting challenge. Here, we describe "X-gene" genetic analysis (XGA), a strategy for engineering and profiling highly combinatorial gene perturbations. We demonstrate XGA on yeast ABC transporters by engineering 5,353 strains, each deleted for a random subset of 16 transporters, and profiling each strain's resistance to 16 compounds. XGA yielded 85,648 genotype-to-resistance observations, revealing high-order genetic interactions for 13 of the 16 transporters studied. Neural networks yielded intuitive functional models and guided exploration of fluconazole resistance, which was influenced non-additively by five genes. Together, our results showed that highly combinatorial genetic perturbation can functionally dissect complex traits, supporting pursuit of analogous strategies in human cells and other model systems.
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
4-(Chloromethyl)benzoyl chloride, 97%
Sigma-Aldrich
Cycloheximide, Biotechnology Performance Certified
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Imatinib
Sigma-Aldrich
Bisantrene dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
Itraconazole, ≥98% (HPLC)
Supelco
Benomyl, PESTANAL®, analytical standard
Sigma-Aldrich
Ketoconazole, 99.0-101.0% (EP, titration)
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Methotrexate, meets USP testing specifications
Sigma-Aldrich
Beauvericin, ≥97% (HPLC)
Sigma-Aldrich
Tamoxifen, ≥99%