Merck
  • Cytochrome P450-derived arachidonic acid metabolism in the rat kidney: characterization of selective inhibitors.
Cytochrome P450-derived arachidonic acid metabolism in the rat kidney: characterization of selective inhibitors.
The Journal of pharmacology and experimental therapeutics (1998-03-13)
M H Wang, E Brand-Schieber, B A Zand, X Nguyen, J R Falck, N Balu, M L Schwartzman
ABSTRACT
We characterized the inhibitory activity of several acetylenic and olefinic compounds on cytochrome P450 (CYP)-derived arachidonic acid omega-hydroxylation and epoxidation using rat renal cortical microsomes and recombinant CYP proteins. Among the acetylenic compounds, 6-(2-propargyloxyphenyl)hexanoic acid (PPOH) and N-methylsulfonyl-6-(2-propargyloxyphenyl)hexanamide were found to be potent and selective inhibitors of microsomal epoxidation with IC50 values of 9 and 13 microM, respectively. On the other hand, 17-octadecynoic acid inhibited both omega-hydroxylation and epoxidation of arachidonic acid with IC50 values of 7 and 5 microM, respectively. The olefinic compounds N-methylsulfonyl-12, 12-dibromododec-11-enamide (DDMS) and 12, 12-dibromododec-11-enoic acid (DBDD) exhibited a high degree of selectivity inhibiting microsomal omega-hydroxylation with an IC50 value of 2 microM, whereas the IC50 values for epoxidation were 60 and 51 microM for DDMS and DBDD, respectively. Studies using recombinant rat CYP4A isoforms showed that PPOH caused a concentration-dependent inhibition of omega-hydroxylation and 11, 12-epoxidation by CYP4A3 or CYP4A2 but had no effect on CYP4A1-catalyzed omega-hydroxylase activity. On the other hand, DDMS inhibited both CYP4A1- and CYP4A3- or CYP4A2-catalyzed arachidonic acid oxidations. Inhibition of microsomal activity by PPOH, but not DDMS, was time- and NADPH-dependent, a result characteristic of a mechanism-based irreversible inhibitor. These studies provide information useful for evaluating the role of the CYP-derived arachidonic acid metabolites in the regulation of renal function and blood pressure.
Sigma-Aldrich
17-Octadecynoic acid, ≥95% (GC)