Merck
  • Monoclonal alkaline phosphatase-anti-alkaline phosphatase (APAAP) complex: production of antibody, optimization of activity, and use in immunostaining.
Monoclonal alkaline phosphatase-anti-alkaline phosphatase (APAAP) complex: production of antibody, optimization of activity, and use in immunostaining.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1988-02-01)
A Hohmann, A J Hodgson, W Di, J M Skinner, J Bradley, H Zola
ABSTRACT
A mouse monoclonal antibody, FMC55 (an IgG1), to alkaline phosphatase was prepared and evaluated in immunostaining. Clones producing antibody to alkaline phosphatase were selected using a micro-ELISA which identified antibodies forming active soluble complexes (APAAP) with the enzyme. Conditions that influenced the formation of the complex were investigated by using a quantitative assay in which the complex was captured by a bridging anti-mouse antibody. The ratio of FMC55 to enzyme had a major influence on the activity of the complex. Although all complexes had some activity, those that contained excess antibody had reduced ability to bind to anti-mouse antibody because of competition with excess unlabeled antibody. The optimal complex was formed with 3 micrograms of FMC55 per unit of enzyme. This complex contained neither free enzyme nor free antibody. The molecular weight by gel permeation chromatography was 600,000, giving a composition of two enzyme and two antibody molecules or one enzyme and three antibody molecules. The size of the complex was not altered by adding excess antibody or excess enzyme. Immunoblotting showed that FMC55 bound only to the Mr 140,000 homodimeric form of alkaline phosphatase. The APAAP complex was used in combination with biotin-streptavidin-peroxidase reagent to detect two antigens labeled with two different mouse monoclonal antibodies in the same tissue preparation.
Sigma-Aldrich
Phosphatase, Alkaline from bovine intestinal mucosa, lyophilized powder, ≥10 DEA units/mg solid