MilliporeSigma
  • A Comparative Study of Inhaled Nitric Oxide and an Intravenously Administered Nitric Oxide Donor in Acute Pulmonary Hypertension.
A Comparative Study of Inhaled Nitric Oxide and an Intravenously Administered Nitric Oxide Donor in Acute Pulmonary Hypertension.
Drug design, development and therapy (2020-02-29)
Anna Stene Hurtsén, Ilya Zorikhin Nilsson, Emanuel M Dogan, Kristofer F Nilsson
ABSTRACT
Inhaled nitric oxide (iNO) selectively vasodilates the pulmonary circulation but the effects are sometimes insufficient. Available intravenous (iv) substances are non-selective and cause systemic side effects. The pulmonary and systemic effects of iNO and an iv mono-organic nitrite (PDNO) were compared in porcine models of acute pulmonary hypertension. In anesthetized piglets, dose-response experiments of iv PDNO at normal pulmonary arterial pressure (n=10) were executed. Dose-response experiments of iv PDNO (n=6) and iNO (n=7) were performed during pharmacologically induced pulmonary hypertension (U46619 iv). The effects of iv PDNO and iNO were also explored in 5 mins of hypoxia-induced increase in pulmonary pressure (n=2-4). PDNO (15, 30, 45 and 60 nmol NO kg-1 min-1 iv) and iNO (5, 10, 20 and 40 ppm which corresponded to 56, 112, 227, 449 nmol NO kg-1 min-1, respectively) significantly decreased the U46619-increased mean pulmonary arterial pressure (MPAP) and pulmonary vascular resistance (PVR) to a similar degree without significant decreases in mean arterial pressure (MAP) or systemic vascular resistance (SVR). iNO caused increased levels of methemoglobin. At an equivalent delivered NO quantity (iNO 5 ppm and PDNO 45 nmol kg-1 min-1 iv), PDNO decreased PVR and SVR significantly more than iNO. Both drugs counteracted hypoxia-induced pulmonary vasoconstriction and they decreased the ratio of PVR and SVR in both settings. Intravenous PDNO was a more potent pulmonary vasodilator than iNO in pulmonary hypertension, with no severe side effects. Hence, this study supports the potential of iv PDNO in the treatment of acute pulmonary hypertension.
Product Description
Supelco
Cefuroxime, VETRANAL®, analytical standard