MilliporeSigma
  • Inhibition of BCL2 Family Members Increases the Efficacy of Copper Chelation in BRAFV600E-Driven Melanoma.
Inhibition of BCL2 Family Members Increases the Efficacy of Copper Chelation in BRAFV600E-Driven Melanoma.
Cancer research (2020-02-02)
Ye-Jin Kim, Tiffany Tsang, Grace R Anderson, Jessica M Posimo, Donita C Brady
ABSTRACT
The principal unmet need in BRAFV600E-positive melanoma is lack of an adequate therapeutic strategy capable of overcoming resistance to clinically approved targeted therapies against oncogenic BRAF and/or the downstream MEK1/2 kinases. We previously discovered that copper (Cu) is required for MEK1 and MEK2 activity through a direct Cu-MEK1/2 interaction. Repurposing the clinical Cu chelator tetrathiomolybdate (TTM) is supported by efficacy in BRAFV600E-driven melanoma models, due in part to inhibition of MEK1/2 kinase activity. However, the antineoplastic activity of Cu chelators is cytostatic. Here, we performed high-throughput small-molecule screens to identify bioactive compounds that synergize with TTM in BRAFV600E-driven melanoma cells. Genetic perturbation or pharmacologic inhibition of specific members of the BCL2 family of antiapoptotic proteins (BCL-W, BCL-XL, and MCL1) selectively reduced cell viability when combined with a Cu chelator and induced CASPASE-dependent cell death. Further, in BRAFV600E-positive melanoma cells evolved to be resistant to BRAF and/or MEK1/2 inhibitors, combined treatment with TTM and the clinically evaluated BCL2 inhibitor, ABT-263, restored tumor growth suppression and induced apoptosis. These findings further support Cu chelation as a therapeutic strategy to target oncogene-dependent tumor cell growth and survival by enhancing Cu chelator efficacy with chemical inducers of apoptosis, especially in the context of refractory or relapsed BRAFV600E-driven melanoma. SIGNIFICANCE: This study unveils a novel collateral drug sensitivity elicited by combining copper chelators and BH3 mimetics for treatment of BRAFV600E mutation-positive melanoma.
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis
Sigma-Aldrich
Boron-11B, 95 atom % 11B