MilliporeSigma
  • Development of a new oral poliovirus vaccine for the eradication end game using codon deoptimization.
Development of a new oral poliovirus vaccine for the eradication end game using codon deoptimization.
NPJ vaccines (2020-03-29)
Jennifer L Konopka-Anstadt, Ray Campagnoli, Annelet Vincent, Jing Shaw, Ling Wei, Nhien T Wynn, Shane E Smithee, Erika Bujaki, Ming Te Yeh, Majid Laassri, Tatiana Zagorodnyaya, Amy J Weiner, Konstantin Chumakov, Raul Andino, Andrew Macadam, Olen Kew, Cara C Burns
ABSTRACT
Enormous progress has been made in global efforts to eradicate poliovirus, using live-attenuated Sabin oral poliovirus vaccine (OPV). However, as the incidence of disease due to wild poliovirus has declined, vaccine-derived poliovirus (VDPV) has emerged in areas of low-vaccine coverage. Coordinated global cessation of routine, type 2 Sabin OPV (OPV2) use has not resulted in fewer VDPV outbreaks, and continued OPV use in outbreak-response campaigns has seeded new emergences in low-coverage areas. The limitations of existing vaccines and current eradication challenges warranted development of more genetically stable OPV strains, most urgently for OPV2. Here, we report using codon deoptimization to further attenuate Sabin OPV2 by changing preferred codons across the capsid to non-preferred, synonymous codons. Additional modifications to the 5' untranslated region stabilized known virulence determinants. Testing of this codon-deoptimized new OPV2 candidate (nOPV2-CD) in cell and animal models demonstrated that nOPV2-CD is highly attenuated, grows sufficiently for vaccine manufacture, is antigenically indistinguishable from Sabin OPV2, induces neutralizing antibodies as effectively as Sabin OPV2, and unlike Sabin OPV2 is genetically stable and maintains an attenuation phenotype. In-human clinical trials of nOPV2-CD are ongoing, with potential for nOPV strains to serve as critical vaccine tools for achieving and maintaining polio eradication.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in sheep, affinity isolated antibody, buffered aqueous solution