MilliporeSigma
  • Expression of activating transcription factor 5 (ATF5) is mediated by microRNA-520b-3p under diverse cellular stress in cancer cells.
Expression of activating transcription factor 5 (ATF5) is mediated by microRNA-520b-3p under diverse cellular stress in cancer cells.
PloS one (2020-07-01)
Kari A Gaither, Christy J W Watson, Bhanupriya Madarampalli, Philip Lazarus
ABSTRACT
Cellular stress response mechanisms normally function to enhance survival and allow for cells to return to homeostasis following an adverse event. Cancer cells often co-opt these same mechanisms as a means to evade apoptosis and mitigate a state of constant cellular stress. Activating transcription factor 5 (ATF5) is upregulated under diverse stress conditions and is overexpressed in a variety of cancers. It was demonstrated ATF5 is a survival factor in transformed, but not normal cells. However, the regulation of ATF5 is not fully understood. The purpose of the present study was to investigate miRNA regulation at the 3' untranslated region (UTR) of ATF5, with the goal of demonstrating a reversal of the upregulation of ATF5 induced under diverse cellular stress in cancer cells. A multifactorial approach using in silico analysis was employed to identify miRNAs 433-3p, 520b-3p, and 129-5p as potential regulators of ATF5, based on their predicted binding sites over the span of the ATF5 3' UTR. Luciferase reporter assay data validated all three miRNA candidates by demonstrating direct binding to the target ATF5 3'. However, functional studies revealed miR-520b-3p as the sole candidate able to reverse the upregulation of ATF5 protein under diverse cellular stress. Additionally, miR-520b-3p levels were inversely related to ATF5 mRNA under endoplasmic reticulum stress and amino acid deprivation. This is the first evidence that regulation at the 3' UTR is involved in modulating ATF5 levels under cellular stress and suggests an important role for miRNA-520b-3p in the regulation of ATF5.
Sigma-Aldrich
Anti-β-Tubulin antibody, Mouse monoclonal, clone D66, purified from hybridoma cell culture
Sigma-Aldrich
Anti-ATF5 antibody produced in rabbit, affinity isolated antibody