MilliporeSigma
  • Sensitive, rapid and easy analysis of three catecholamine metabolites in human urine and serum by liquid chromatography tandem mass spectrometry.
Sensitive, rapid and easy analysis of three catecholamine metabolites in human urine and serum by liquid chromatography tandem mass spectrometry.
Journal of chromatographic science (2012-04-19)
Lanyun Fang, Yan Lv, Xuefei Sheng, Shanshan Yao
ABSTRACT
A sensitive and easy analytical method for catecholamine metabolites including 4-hydroxy-3-methoxyphenylglycol sulfate (HMPG sulfate), vanillylmandelic acid (VMA) and homovanillic acid (HVA) determination was developed based on liquid chromatography-tandem mass spectrometry in a negative multiple reaction monitoring mode. The analytes were rapidly separated on a reversed-phase Waters Xbridge C18 column (150 × 2.1 mm i.d.) with the mobile phase of 15% (v/v) acetonitrile containing 2 mM ammonium formate and 85% (v/v) formic acid solution (0.05%, v/v). Mass spectrometric conditions, such as characteristic fragmentations and quantification ion transitions, both with chromatographic conditions including separation column type and mobile phase composition, were systematically investigated to get optimal sensitivity and specificity. The limits of detection were in the range of 0.03-0.7 ng/mL for the targets. Recovery rates of spiked urine samples with three different concentration levels (low, middle and high) were above 86% with precisions less than 5.7%. For serum analysis, acetonitrile chosen both as protein precipitation reagent and extraction solvent facilitates to reduce matrix effects. Recovery rates of spiked serum sample were in the range of 90.6% to 111.1% for three targets. The intra-day and inter-day precisions were satisfactory less than 8.7%. This proposed method was successfully applied to determine HMPG sulfate, HVA and VMA present in human urine and serum.
Sigma-Aldrich
DL-4-Hydroxy-3-methoxymandelic acid, ≥98% (HPLC), powder
Sigma-Aldrich
Formic acid solution, BioUltra, 1.0 M in H2O