MilliporeSigma
  • Improved serodiagnosis of histoplasmosis by use of deglycosylated extracellular released antigens of Histoplasma capsulatum.
Improved serodiagnosis of histoplasmosis by use of deglycosylated extracellular released antigens of Histoplasma capsulatum.
Journal of microbiological methods (2020-06-15)
Primavera Alvarado, Yenis Pérez-Rojas, Edgar Armando Zambrano, Mary I Gonzatti, Antonio Roschman-González
ABSTRACT
The diagnosis of histoplasmosis depends on various approaches: direct clinical examination, fungus isolation from cultures of clinical samples, histopathological evaluation, and serological testing. In serodiagnostic assays, the Histoplasma capsulatum H and M antigenic glycoproteins have been extensively used. However, both antigens showed limitations attributed mainly to their cross-reactivity with glycoproteins from other pathogenic fungi, which compromises specificity, and generates false positives, misdiagnosis, and therapeutic failure. In this work, we deglycosylated extracellular released antigens from the Venezuelan 7090 H. capsulatum clinical isolate, using chemical and enzymatic methods and evaluated their effectiveness by indirect enzyme-linked immunosorbent assay (ELISA) with sera from patients with either histoplasmosis or PCM. Prior to deglycosylation, the extracellular released antigen showed 62% of sensitivity 66% of specificity and 68% of cross-reactivity with paracoccidioidomicosis sera. The chemically deglycosylated extracellular released antigen, for 8 or 18  h showed 72 and 52% sensitivity with 98% and 92% specificity, respectively. Moreover, cross-reactivity with Paracoccidioides decreased to 4 and 16%, following deglycosylation for 8 or 18 h, respectively. The enzymatically treated antigen showed 52% of sensitivity, 92% of specificity and 8% cross-reactivity against Paracoccidioides. Deglycosylation of the H. capsulatum antigen improves its specificity and decreases its cross-reactivity against Paracoccidioides when using indirect ELISA for serodiagnosis. Therefore, it is recommended to deglycosylate the fungal extracellular released antigen for clinical serodiagnosis, and to monitor humoral immune responses during therapy of patients with the different clinical forms of histoplasmosis.
Sigma-Aldrich
Enzymatic Protein Deglycosylation Kit