MilliporeSigma
  • Red and blue light differentially impact retrograde signalling and photoprotection in rice.
Red and blue light differentially impact retrograde signalling and photoprotection in rice.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences (2020-05-05)
Liu Duan, M Águila Ruiz-Sola, Ana Couso, Nil Veciana, Elena Monte
ABSTRACT
Chloroplast-to-nucleus retrograde signalling (RS) is known to impact plant growth and development. In Arabidopsis, we and others have shown that RS affects seedling establishment by inhibiting deetiolation. In the presence of lincomycin, a chloroplast protein synthesis inhibitor that triggers RS, Arabidopsis light-grown seedlings display partial skotomorphogenesis with undeveloped plastids and closed cotyledons. By contrast, RS in monocotyledonous has been much less studied. Here, we show that emerging rice seedlings exposed to lincomycin do not accumulate chlorophyll but otherwise remain remarkably unaffected. However, by using high red (R) and blue (B) monochromatic lights in combination with lincomycin, we have uncovered a RS inhibition of length and a reduction in the B light-induced declination of the second leaf. Furthermore, we present data showing that seedlings grown in high B and R light display different non-photochemical quenching capacity. Our findings support the view that excess B and R light impact seedling photomorphogenesis differently to photoprotect and optimize the response to high-light stress. This article is part of the theme issue 'Retrograde signalling from endosymbiotic organelles'.
Sigma-Aldrich
Lincomycin hydrochloride, BioReagent, suitable for cell culture