MilliporeSigma
  • Inhibition of JNJ-26481585-mediated autophagy induces apoptosis via ROS activation and mitochondrial membrane potential disruption in neuroblastoma cells.
Inhibition of JNJ-26481585-mediated autophagy induces apoptosis via ROS activation and mitochondrial membrane potential disruption in neuroblastoma cells.
Molecular and cellular biochemistry (2020-03-09)
Vamsi Krishna Kommalapati, Dinesh Kumar, Anjana Devi Tangutur
ABSTRACT
Neuroblastoma (NB) is the common pediatric tumor of the sympathetic nervous system characterized by poor prognosis. Owing to the challenges such as high tumor heterogeneity, multidrug resistance, minimal residual disease, etc., there is an immediate need for exploring new therapeutic strategies and effective treatments for NB. Herein, in the current study, we explored the unexplored response of NB cells to the second-generation histone deacetylase inhibitor (HDACi) JNJ-26481585(JNJ) and the lysosomotropic agent, Chloroquine (CQ) alone and upon JNJ/CQ treatment as a plausible therapeutic. We identify that while JNJ alone induced autophagy in NB cells, JNJ/CQ treatment decreased the viability and proliferation of NB cells in vitro by switching from autophagy to apoptosis. Further we found that autophagy inhibition by CQ pre-treatment led to the generation of ROS and a decrease in the mitochondrial membrane potential (MMP) that subsequently caused caspase-3-mediated apoptotic cell death in NB cells. Corroborating the above observations, we found that the ROS scavenger N-acetylcysteine (NAC) countered caspase-3 activity and the cells were rescued from apoptosis. Finally, these observations establish that JNJ/CQ treatment resulted in cell death in NB cells by triggering the formation of ROS and disruption of MMP, suggesting that modulation of JNJ-induced autophagy by CQ represents a promising new therapeutic approach in NB.
Sigma-Aldrich
Dansylcadaverine, suitable for fluorescence, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Z-VAD-FMK, solid
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
Acridine Orange hemi(zinc chloride) salt, For nucleic acid staining in cells or gels
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium - high glucose, With 4500 mg/L glucose, L-glutamine, and sodium pyruvate, without sodium bicarbonate, powder, suitable for cell culture
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)