Merck
  • A cyclic dipeptide from the Chilean hazelnut cotyledons (Gevuina avellana Mol., Proteaceae).
A cyclic dipeptide from the Chilean hazelnut cotyledons (Gevuina avellana Mol., Proteaceae).
Scientific reports (2020-04-29)
Guillermo Schmeda-Hirschmann, Jean Paulo de Andrade, Felipe Jiménez-Aspee, Daniel Mieres-Castro
ABSTRACT
The Chilean hazelnut (Gevuina avellana Mol., Proteaceae) is a southern South American nut consumed as a snack and included in different preparations of traditional Chilean cuisine. Recently we described the fatty acid profile, oxylipins, phenolic compounds, as well as the antioxidant capacity. The main compounds of the phenolic-enriched extract were only tentatively identified by spectrometric means. In the present work, we describe the isolation and full characterization of a cyclic dipeptide cyclo(Arg-Trp) and other compounds from the phenolic enriched extracts of the G. avellana cotyledons. Compounds were isolated by means of counter-current chromatography and structures were established by spectroscopic and spectrometric methods. This is the first report on small peptides in G. avellana and adds evidence on the possible beneficial effects of this nut in human health.
Product Description
Supelco
Amberlite XAD7HP, 20-60 mesh