MilliporeSigma
  • The Hippo pathway oncoprotein YAP promotes melanoma cell invasion and spontaneous metastasis.
The Hippo pathway oncoprotein YAP promotes melanoma cell invasion and spontaneous metastasis.
Oncogene (2020-06-21)
Xiaomeng Zhang, Lie Yang, Pacman Szeto, Gamze Kuser Abali, Youfang Zhang, Aishwarya Kulkarni, Kaushalya Amarasinghe, Jason Li, Ismael A Vergara, Ramyar Molania, Anthony T Papenfuss, Catriona McLean, Mark Shackleton, Kieran F Harvey
ABSTRACT
Melanoma is a deadly form of skin cancer that accounts for a disproportionally large proportion of cancer-related deaths in younger people. Compared with most other skin cancers, a feature of melanoma is its high metastatic capacity, although the mechanisms that confer this are not well understood. The Hippo pathway is a key regulator of organ growth and cell fate that is deregulated in many cancers. To analyse the Hippo pathway in cutaneous melanoma, we generated a transcriptional signature of melanoma cells that overexpressed YAP, the key downstream Hippo pathway oncoprotein. YAP-mediated transcriptional activity varied in melanoma cell lines but did not cluster with known genetic drivers of melanomagenesis such as BRAF and NRAS mutations. Instead, it correlated strongly with published gene expression profiles linked to melanoma cell invasiveness and varied throughout the metastatic cascade in melanoma patient tumours. Consistent with this, YAP was both necessary and sufficient for melanoma cell invasion in vitro. In vivo, YAP promoted spontaneous melanoma metastasis, whilst the growth of YAP-expressing primary tumours was impeded. Finally, we identified the YAP target genes AXL, THBS1 and CYR61 as key mediators of YAP-induced melanoma cell invasion. These data suggest that YAP is a critical regulator of melanoma metastasis.
Sigma-Aldrich
Anti-CRIM1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Phalloidin-Atto 490LS, suitable for fluorescence