MilliporeSigma
Versatile 3' Functionalization of CRISPR Single Guide RNA.
Chembiochem : a European journal of chemical biology (2020-01-17)
Cody M Palumbo, Jeton M Gutierrez-Bujari, Henriette O'Geen, David J Segal, Peter A Beal
ABSTRACT
Specific applications of CRISPR/Cas genome editing systems benefit from chemical modifications of the sgRNA. Herein we describe a versatile and efficient strategy for functionalization of the 3'-end of a sgRNA. An exemplary collection of six chemically modified sgRNAs was prepared containing crosslinkers, a fluorophore and biotin. Modification of the sgRNA 3'-end was broadly tolerated by Streptococcus pyogenes Cas9 in an in vitro DNA cleavage assay. The 3'-biotinylated sgRNA was used as an affinity reagent to identify IGF2BP1, YB1 and hnRNP K as sgRNA-binding proteins present in HEK293T cells. Overall, the modification strategy presented here has the potential to expand on current applications of CRISPR/Cas systems.
Sigma-Aldrich
Iodoacetic acid N-hydroxysuccinimide ester, powder