MilliporeSigma
  • Aconitine induces cardiotoxicity through regulation of calcium signaling pathway in zebrafish embryos and in H9c2 cells.
Aconitine induces cardiotoxicity through regulation of calcium signaling pathway in zebrafish embryos and in H9c2 cells.
Journal of applied toxicology : JAT (2020-01-25)
Mengting Li, Xiaofang Xie, Haimei Chen, Qiuyun Xiong, Rongsheng Tong, Cheng Peng, Fu Peng
ABSTRACT
Fuzi, the processed lateral roots of Aconitum carmichaelii Debx., is a traditional herbal medicine that is well known for its excellent pharmacological effects and acute toxicity. Aconitine is one of the diester-diterpene alkaloids and well-known for its arrhythmogenic effects. However, the effects of aconitine in zebrafish have rarely been studied. Therefore, we investigated the effects of aconitine on zebrafish embryos and H9c2 cells. Zebrafish embryos at 48 hours postfertilization were exposed to aconitine, and then, cardiac function and apoptosis were measured. Through transcriptomic analysis, the cardiotoxicity of aconitine in zebrafish embryos was involved in regulating Ca2+ signal pathways. A reverse transcription-polymerase chain reaction was performed to verify the expression of Ca2+ pathway-related genes after 12, 24, 36 and 48 hours of treatment. Meanwhile, intracellular Ca2+ concentrations and cell apoptosis were observed in H9c2 cells treated with half-maximal inhibitory concentration values of aconitine for 30 minutes. The protein levels of troponin T (TnT), caspase 3, Bcl-2 and Bax were detected by western blot analysis. In vivo, 2.0 and 8.0 μm aconitine decreased the heart rate and inhibited the contraction of ventricles and atria in a dose- and time-dependent manner. Furthermore, aconitine increased expression of cacna1c, RYR2, atp2a2b, Myh6, troponin C, p38, caspase 3, Bcl-2 and Bax for 12 hours. In vitro, 1.5 and 4.5 mm aconitine caused intracellular Ca2+ ion oscillation, increased rates of apoptosis, inhibited TnT and Bcl-2 protein expression, and promoted caspase 3 and Bax protein expression. These data confirmed that aconitine at various concentrations induced cardiac dysfunction and apoptosis were related to the Ca2+ signaling pathway.
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP