MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Pegylated Liposomal Doxorubicin/Oxaliplatin Chemotherapy Can Overcome Cisplatin Resistance in Spectrin αII-Overexpressing Ovarian Carcinoma.
Pegylated Liposomal Doxorubicin/Oxaliplatin Chemotherapy Can Overcome Cisplatin Resistance in Spectrin αII-Overexpressing Ovarian Carcinoma.
Anticancer research (2020-05-06)
Osamu Maeda, Hiroaki Kajiyama, Kiyosumi Shibata, Shigeo Nakamura, Fumitaka Kikkawa
ABSTRACT
Spectrin αII contributes to cisplatin and carboplatin resistance in ovarian serous carcinoma cells, and its expression in surgical specimens is a valid predictor of prognosis. We sought to identify effective drugs for spectrin αII-mediated cisplatin-resistant cells. We employed SKOV3 cells with small interfering RNA-mediated spectrin αII downregulation, serous carcinoma cells (NOS2), cisplatin-resistant cells (NOS2CR2), and oxaliplatin-resistant cells (NOS2OXR). In the drug-sensitivity test, oxaliplatin was not affected by the inhibition of spectrin αII expression and was effective for cisplatin-resistant NOS2CR2 cells. NOS2OXR cells did not express higher levels of spectrin αII compared to NOS2 in western blot analysis. Six non-platinum anticancer drugs were not affected by the inhibition and was effective for resistant NOS2CR2 and NOS2OXR cells. Doxorubicin exhibited potent cytotoxicity at 2 μM against both resistant cell lines. Pegylated liposomal doxorubicin/oxaliplatin regimen may be effective for cisplatin-resistant ovarian carcinoma with spectrin αII-overexpression.
Product Description
BRAND® 96-well microplate, U-bottom, round bottom, non-sterile