MilliporeSigma
  • Applicability of Supercritical fluid chromatography-Mass spectrometry to metabolomics. II-Assessment of a comprehensive library of metabolites and evaluation of biological matrices.
Applicability of Supercritical fluid chromatography-Mass spectrometry to metabolomics. II-Assessment of a comprehensive library of metabolites and evaluation of biological matrices.
Journal of chromatography. A (2020-03-18)
Gioacchino Luca Losacco, Omar Ismail, Julian Pezzatti, Víctor González-Ruiz, Julien Boccard, Serge Rudaz, Jean-Luc Veuthey, Davy Guillarme
ABSTRACT
In this work, the impact of biological matrices, such as plasma and urine, was evaluated under SFCHRMS in the field of metabolomics. For this purpose, a representative set of 49 metabolites were selected. The assessment of the matrix effects (ME), the impact of biological fluids on the quality of MS/MS spectra and the robustness of the SFCHRMS method were each taken into consideration. The results have highlighted a limited presence of ME in both plasma and urine, with 30% of the metabolites suffering from ME in plasma and 25% in urine, demonstrating a limited sensitivity loss in the presence of matrices. Subsequently, the MS/MS spectra evaluation was performed for further peak annotation. Their analyses have highlighted three different scenarios: 63% of the tested metabolites did not suffer from any interference regardless of the matrix; 21% were negatively impacted in only one matrix and the remaining 16% showed the presence of matrix-belonging compounds interfering in both urine and plasma. Finally, the assessment of retention times stability in the biological samples, has brought into evidence a remarkable robustness of the SFCHRMS method. Average RSD (%) values of retention times for spiked metabolites were equal or below 0.5%, in the two biological fluids over a period of three weeks. In the second part of the work, the evaluation of the Sigma Mass Spectrometry Metabolite Library of Standards containing 597 metabolites, under SFCHRMS conditions was performed. A total detectability of the commercial library up to 66% was reached. Among the families of detected metabolites, large percentages were met for some of them. Highly polar metabolites such as amino acids (87%), nucleosides (85%) and carbohydrates (71%) have demonstrated important success rates, equally for hydrophobic analytes such as steroids (78%) and lipids (71%). On the negative side, very poor performance was found for phosphorylated metabolites, namely phosphate-containing compounds (14%) and nucleotides (31%).
Sigma-Aldrich
Mass Spectrometry Metabolite Library, Supplied by IROA Technologies