MilliporeSigma
  • Specific IgG for cat allergens in patients with allergic conjunctivitis.
Specific IgG for cat allergens in patients with allergic conjunctivitis.
International ophthalmology (2014-09-06)
Anri Miyama, Tatsuya Mimura, Hidetaka Noma, Mari Goto, Yuko Kamei, Aki Kondo, Yusuke Saito, Hiroko Okuma, Masao Matsubara
ABSTRACT
Immunoglobulin G (IgG) antibodies are involved in type II and type III hypersensitivity. We evaluated the relation between perennial allergic conjunctivitis and serum levels of specific IgG for cat allergens. A prospective study was conducted in patients with seasonal allergic conjunctivitis (seasonal group, n = 10), patients with perennial allergic conjunctivitis (perennial group, n = 10), and healthy control subjects (control group, n = 10). Serum levels of specific IgE and IgG for cat allergens and total tear IgE were measured, and a skin prick test was also performed. In addition, a severity score associated with allergic conjunctivitis was calculated (0-30). The positive rates and scores of for total tear IgE, serum cat-specific IgE, and serum cat-specific IgG were all higher in the seasonal and perennial groups than in the control group (all p < 0.05). Serum cat-specific IgG levels were higher in the perennial group than in the seasonal group (p = 0.0156), but there was no significant difference in the grade of cat-specific IgE between the two groups (p = 0.3008). On multivariate analysis, the mean wheal diameter for cat allergen was associated with the serum level of cat-specific IgG (not IgE) in all patients [odds ratio (OR) = 31.979, p < 0.0001]. Multivariate analysis revealed that the total objective score was strongly associated with serum cat-specific IgG (OR = 23.015, p < 0.0001). These findings suggest that specific IgG antibodies may be involved in perennial allergic symptoms caused by indoor allergens such as cat allergens.