MilliporeSigma
  • Inhibitory effect of (-)-epigallocatechin and (-)-epigallocatechin gallate against heregulin beta1-induced migration/invasion of the MCF-7 breast carcinoma cell line.
Inhibitory effect of (-)-epigallocatechin and (-)-epigallocatechin gallate against heregulin beta1-induced migration/invasion of the MCF-7 breast carcinoma cell line.
Biological & pharmaceutical bulletin (2009-05-08)
Yukihiro Kushima, Kazuki Iida, Yasuo Nagaoka, Yasuyuki Kawaratani, Tatsuya Shirahama, Minoru Sakaguchi, Kimiye Baba, Yukihiko Hara, Shinichi Uesato
ABSTRACT
Migration/invasion is involved in the multiple steps of metastasis, resulting in a poor prognosis of breast cancer. (-)-Epigallocatechin-3-gallate (EGCG) in green tea inhibits the metastasis of some cancer cell lines. Cell migration/invasion assays using Boyden chambers demonstrated that (-)-epigallocatechin (EGC), another green tea catechin, inhibited heregulin-beta1 (HRG)-induced migration/invasion of MCF-7 human breast carcinoma cells to approximately the same extent as EGCG. Assays of cytoskeletal reorganization, Western blotting and immunoprecipitation suggested that EGCG inhibited this migration/invasion by suppressing the HRG-stimulated activation of epidermal growth factor receptor-related protein B2 (ErbB2)/ErbB3/protein kinase B (Akt), whereas EGC did so through pathways including the disruption of the HRG-stimulated activation of ErbB2/ErbB3 but not Akt. EGC, as well as EGCG, could play an important role against the promotion of metastasis of breast cancer cells.
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 mg
In Stock
Details...
$139.00