MilliporeSigma
  • Melatonin inhibits aromatase promoter expression by regulating cyclooxygenases expression and activity in breast cancer cells.
Melatonin inhibits aromatase promoter expression by regulating cyclooxygenases expression and activity in breast cancer cells.
British journal of cancer (2009-09-24)
C Martínez-Campa, A González, M D Mediavilla, C Alonso-González, V Alvarez-García, E J Sánchez-Barceló, S Cos
ABSTRACT
Melatonin reduces the development of breast cancer interfering with oestrogen-signalling pathways, and also inhibits aromatase activity and expression. Our objective was to study the promoters through which melatonin modifies aromatase expression, evaluate the ability of melatonin to regulate cyclooxygenases and assess whether the effects of melatonin are related to its effects on intracellular cAMP, in MCF-7 cells. Total aromatase mRNA, aromatase mRNA promoter regions and cyclooxygenases mRNA expression were determined by real-time RT-PCR. PGE(2) and cAMP were measured by kits. Melatonin downregulated the gene expression of the two major specific aromatase promoter regions, pII and pI.3, and also that of the aromatase promoter region pI.4. Melatonin 1 nM was able to counteract the stimulatory effect of tetradecanoyl phorbol acetate on PGE(2) production and inhibit COX-2 and COX-1 mRNA expression. Melatonin 1 nM elicited a parallel time-dependent decrease in both cyclic AMP formation and aromatase mRNA expression. This study shows that melatonin inhibits aromatase activity and expression by regulating the gene expression of specific aromatase promoter regions. A possible mechanism for these effects would be the regulation by melatonin of intracellular cAMP levels, mediated by an inhibition of cyclooxygenase activity and expression.
Sigma-Aldrich
8-Bromoadenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Luzindole, ≥90%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)