Merck
  • Increased inflammation, impaired bacterial clearance, and metabolic disruption after gram-negative sepsis in Mkp-1-deficient mice.
Increased inflammation, impaired bacterial clearance, and metabolic disruption after gram-negative sepsis in Mkp-1-deficient mice.
Journal of immunology (Baltimore, Md. : 1950) (2009-11-06)
W Joshua Frazier, Xianxi Wang, Lyn M Wancket, Xiang-An Li, Xiaomei Meng, Leif D Nelin, Andrew C B Cato, Yusen Liu
ABSTRACT
MAPKs are crucial for TNF-alpha and IL-6 production by innate immune cells in response to TLR ligands. MAPK phosphatase 1 (Mkp-1) deactivates p38 and JNK, abrogating the inflammatory response. We have previously demonstrated that Mkp-1(-/-) mice exhibit exacerbated inflammatory cytokine production and increased mortality in response to challenge with LPS and heat-killed Staphylococcus aureus. However, the function of Mkp-1 in host defense during live Gram-negative bacterial infection remains unclear. We challenged Mkp-1(+/+) and Mkp-1(-/-) mice with live Escherichia coli i.v. to examine the effects of Mkp-1 deficiency on animal survival, bacterial clearance, metabolic activity, and cytokine production. We found that Mkp-1 deficiency predisposed animals to accelerated mortality and was associated with more robust production of TNF-alpha, IL-6 and IL-10, greater bacterial burden, altered cyclooxygenase-2 and iNOS expression, and substantial changes in the mobilization of energy stores. Likewise, knockout of Mkp-1 also sensitized mice to sepsis caused by cecal ligation and puncture. IL-10 inhibition by neutralizing Ab or genetic deletion alleviated increased bacterial burden. Treatment with the bactericidal antibiotic gentamicin, given 3 h after Escherichia coli infection, protected Mkp-1(+/+) mice from septic shock but had no effect on Mkp-1(-/-) mice. Thus, during Gram-negative bacterial sepsis Mkp-1 not only plays a critical role in the regulation of cytokine production but also orchestrates the bactericidal activities of the innate immune system and controls the metabolic response to stress.
Sigma-Aldrich
Gentamicin sulfate, meets USP testing specifications, powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Gentamicin sulfate, meets EP, USP testing specifications