MilliporeSigma
  • Bicarbonate suppresses mitochondrial membrane depolarization induced by conventional uncouplers.
Bicarbonate suppresses mitochondrial membrane depolarization induced by conventional uncouplers.
Biochemical and biophysical research communications (2020-08-24)
Ljudmila S Khailova, Tatyana V Vygodina, Galina Y Lomakina, Elena A Kotova, Yuri N Antonenko
ABSTRACT
Bicarbonate has been known to modulate activities of various mitochondrial enzymes such as ATPase and soluble adenylyl cyclase. Here, we found that the ability of conventional protonophoric uncouplers, such as 2,4-dinitrophenol (DNP), carbonylcyanide p-trifluoromethoxyphenylhydrazone (FCCP) and carbonyl cyanide m-chlorophenyl hydrazone (CCCP), but not that of the new popular uncoupler BAM15, to decrease mitochondrial membrane potential was significantly diminished in the presence of millimolar concentrations of bicarbonate. Thus, the depolarizing activity of DNP and FCCP in mitochondria could be sensitive to the local concentration of bicarbonate in cells and tissues. However, bicarbonate could not restore the ATP synthesis suppressed by DNP or CCCP in mitochondria. Bicarbonate neither altered the depolarizing action of DNP and FCCP on proteoliposomes with reconstituted cytochrome c oxidase, nor affected the protonophoric activity of DNP and FCCP in artificial lipid membranes as measured with pyranine-loaded liposomes, thereby showing that the bicarbonate-induced reversal of the depolarizing action of DNP and FCCP on mitochondria did not result from direct interaction of bicarbonate with the uncouplers.
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Tyrphostin A9, solid
Sigma-Aldrich
Carbonyl cyanide 3-chlorophenylhydrazone, ≥97% (TLC), powder