MilliporeSigma
  • Enzyme-digested Colla Corii Asini (E'jiao) accelerates wound healing and prevents ultraviolet A-induced collagen synthesis decline and wrinkle formation in three-dimensional skin equivalents.
Enzyme-digested Colla Corii Asini (E'jiao) accelerates wound healing and prevents ultraviolet A-induced collagen synthesis decline and wrinkle formation in three-dimensional skin equivalents.
Human cell (2020-08-08)
Li Xiao, Feng Liao, Yumei Fan, Nobuhiko Miwa
ABSTRACT
Cutaneous wound healing delay, collagen synthesis decline and wrinkle formation are the common features of skin aging. The aim of this study is to investigate repressive effects of Colla Corii Asini (CCA) (a traditional Chinese medicine which has been used for anti-aging) on hydrogen peroxide (300 µM, 2 h) and ultraviolet A (UVA) (3.2 mJ/cm2)-induced skin aging in vitro. To simulate the in vivo condition of CCA, CCA was digested by gastrointestinal enzymes and added to human gingival fibroblasts (HGF) and three dimensional (3D) skin equivalents at different concentrations. Cell viability assay showed that the enzyme-digested CCA (CCAD) exhibited significant preventive effects on hydrogen peroxide- and UVA-induced cell death. The in vitro scratch assay showed that CCAD was able to prevent hydrogen peroxide-induced wound healing delay in HGF cell sheets. Immunostaining and imaging analysis showed that CCAD could suppress UVA-reduced expression of type IV collagen and elastin in both HGF cells and the 3D skin equivalents. Using a tissue stretching system, wrinkles were formed on UVA-irradiated 3D skin equivalents. Without CCAD-treatment, the wrinkles on the skin were deep, whereas CCAD markedly reduced the depth of wrinkles. In conclusion, CCAD could protect skin cells from oxidative stress and UVA-induced harmful effects, accelerate wound healing, promote synthesis of collagen and elastin, and reduce wrinkles formation. CCAD might be developed as an anti-skin aging reagent in the cosmetic industry.
Sigma-Aldrich
2-O-a-D-Glucopyranosyl-L-ascorbic Acid, ≥98% (HPLC)