MilliporeSigma
  • Upregulation of a marine fungal biosynthetic gene cluster by an endobacterial symbiont.
Upregulation of a marine fungal biosynthetic gene cluster by an endobacterial symbiont.
Communications biology (2020-09-25)
Mingwei Shao, Changli Sun, Xiaoxiao Liu, Xiaoxue Wang, Wenli Li, Xiaoyi Wei, Qinglian Li, Jianhua Ju
ABSTRACT
Fungal-bacterial associations are present in nature, playing important roles in ecological, evolutionary and medicinal processes. Here we report a fungus-bacterial symbiont from marine sediment. The bacterium lives inside the fungal mycelium yet is robust enough to survive independent of its host; the independently grown bacterium can infect the fungal host in vitro and continue to grow progenitively. The bacterial symbiont modulates the fungal host to biosynthesize a polyketide antimicrobial, spiromarmycin. Spiromarmycin appears to endow upon the symbiont pair a protective/defensive means of warding off competitor organisms, be they prokaryotic or eukaryotic microorganisms. Genomic analyses revealed the spiromarmycin biosynthetic machinery to be encoded, not by the bacterium, but rather the fungal host. This unique fungal-bacterial symbiotic relationship and the molecule/s resulting from it dramatically expand our knowledge of marine microbial diversity and shed important insights into endosymbionts and fungal-bacterial relationships.