Merck
  • Exploring the possibilities of infrared spectroscopy for urine sediment examination and detection of pathogenic bacteria in urinary tract infections.
Exploring the possibilities of infrared spectroscopy for urine sediment examination and detection of pathogenic bacteria in urinary tract infections.
Clinical chemistry and laboratory medicine (2020-07-11)
Mieke Steenbeke, Sander De Bruyne, Jerina Boelens, Matthijs Oyaert, Griet Glorieux, Wim Van Biesen, Jere Linjala, Joris R Delanghe, Marijn M Speeckaert
ABSTRACT
Objectives In this study, the possibilities of Fourier-transformed infrared spectroscopy (FTIR) for analysis of urine sediments and for detection of bacteria causing urinary tract infections (UTIs) were investigated. Methods Dried urine specimens of control subjects and patients presenting with various nephrological and urological conditions were analysed using mid-infrared spectroscopy (4,000-400 cm-1). Urine samples from patients with a UTI were inoculated on a blood agar plate. After drying of the pure bacterial colonies, FTIR was applied and compared with the results obtained by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOF MS). Chemometric data analysis was used to classify the different species. Results Due to the typical molecular assignments of lipids, proteins, nucleic acids and carbohydrates, FTIR was able to identify bacteria and showed promising results in the detection of proteins, lipids, white and red blood cells, as well as in the identification of crystals. Principal component analysis (PCA) allowed to differentiate between Gram-negative and Gram-positive species and soft independent modelling of class analogy (SIMCA) revealed promising classification ratios between the different pathogens. Conclusions FTIR can be considered as a supplementary method for urine sediment examination and for detection of pathogenic bacteria in UTI.
Sigma-Aldrich
Hemoglobin human, lyophilized powder