MilliporeSigma
  • Assessment of in vitro and in vivo activities in the National Cancer Institute's anticancer screen with respect to chemical structure, target specificity, and mechanism of action.
Assessment of in vitro and in vivo activities in the National Cancer Institute's anticancer screen with respect to chemical structure, target specificity, and mechanism of action.
Journal of medicinal chemistry (2006-03-17)
Ruili Huang, Anders Wallqvist, David G Covell
ABSTRACT
This paper examines two biological models of anticancer activity, cytotoxicity and hollow fiber (HF) activity, for chemotherapeutic agents evaluated as part of the National Cancer Institute's (NCI's) drug screening effort. Our analysis proposes strategies to globally assess compounds tested in the NCI's 60-cell (NCI60) in vitro anticancer screen in terms of structural features, biological activity, target specificity, and mechanism of action by data integration via our self-organizing maps of structural and biological response patterns. We have built statistical models to predict compound potency and HF activity based on physicochemical properties. Our results find that it is the combination of different structural properties that determines a compound's biological activity. A direct correlation is also found between compound potency and specificity, indicating that specific targeting, rather than promiscuous poisoning, gives rise to potency. Finally, we offer a strategy to exploit this relationship for future mining of novel anticancer candidates.
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mL
In Stock
Details...
$589.00
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mL
In Stock
Details...
$589.00
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mL
In Stock
Details...
$589.00