MilliporeSigma
  • Inhibition of nitric oxide production of activated mice peritoneal macrophages is independent of the Toxoplasma gondii strain.
Inhibition of nitric oxide production of activated mice peritoneal macrophages is independent of the Toxoplasma gondii strain.
Memorias do Instituto Oswaldo Cruz (2021-03-18)
João Cláudio Damasceno-Sá, Fernanda Silva de Souza, Thiago Alves Teixeira Dos Santos, Fábio Conceição de Oliveira, Maria de Fátima Sarro da Silva, Raul Ramos Furtado Dias, Wanderley de Souza, Andrea Cristina Veto Arnholdt, Sergio Henrique Seabra, Renato Augusto DaMatta
ABSTRACT
Toxoplasma gondii causes toxoplasmosis and is controlled by activated macrophages. However, infection of macrophages by tachyzoites induces TGF-β signaling (TGF-s) inhibiting nitric oxide (NO) production. NO inhibition may be a general escape mechanism of distinct T. gondii strains. To evaluate in activated macrophages the capacity of T. gondii strains of different virulence and genetics (RH, type I; ME-49, type II; VEG, type III; P-Br, recombinant) to evade the NO microbicidal defense system and determine LC3 loading to the parasitophorous vacuole. Activated peritoneal macrophages were infected with the different T. gondii strains, NO-production was evaluated by the Griess reagent, and inducible nitric oxide synthase expression, TGF-s, and LC3 localisation assayed by immunofluorescence. Only RH persisted in macrophages, while VEG was more resistant than P-Br and ME-49. All strains induced TGF-s, degradation of inducible nitric oxide synthase, and NO-production inhibition from 2 to 24 h of infection, but only RH sustained these alterations for 48 h. By 24 h of infection, TGF-s lowered in macrophages infected by ME-49, and P-Br, and NO-production recovered, while VEG sustained TGF-s and NO-production inhibition longer. LC3 loading to parasitophorous vacuole was strain-dependent: higher for ME-49, P-Br and VEG, lower for RH. All strains inhibited NO-production, but only RH sustained this effect probably because it persisted in macrophages due to additional evasive mechanisms as lower LC3 loading to parasitophorous vacuole. These results support that T. gondii can escape the NO microbicidal defense system at the initial phase of the infection, but only the virulent strain sustain this evasion mechanism.
Sigma-Aldrich
Anti-LC3A antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution