MilliporeSigma
  • Human INT6 interacts with MCM7 and regulates its stability during S phase of the cell cycle.
Human INT6 interacts with MCM7 and regulates its stability during S phase of the cell cycle.
Oncogene (2007-02-22)
S Buchsbaum, C Morris, V Bochard, P Jalinot
ABSTRACT
The mouse int6 gene is a frequent integration site of the mouse mammary tumor virus and INT6 silencing by RNA interference in HeLa cells causes an increased number of cells in the G2/M phases of the cell cycle, along with mitotic defects. In this report, we investigated the functional significance of the interaction between INT6 and MCM7, which was observed in a two-hybrid screen performed with INT6 as bait. It was found that proteasome inhibition strengthens interaction between both proteins and that INT6 stabilizes MCM7. Removal of MCM7 from chromatin as replication proceeds was accelerated in INT6-silenced cells and reduced amounts of protein were transiently observed, followed by a correction resulting from stimulation of mcm7 gene expression. Synchronized cells depleted for either INT6 or MCM7 display a reduction in thymidine incorporation and a reinforced association of RPA and claspin with chromatin. These data show that INT6 stabilizes chromatin-bound MCM7 and that alteration of this effect is associated with replication deficiency.
Sigma-Aldrich
Anti-Replication Protein A (Ab-1) Mouse mAb (RPA70-9), liquid, clone RPA70-9, Calbiochem®