MilliporeSigma
Influence of limonin on Wnt signalling molecule in HepG2 cell lines.
Journal of natural science, biology, and medicine (2013-05-02)
K Langeswaran, S Gowthamkumar, S Vijayaprakash, R Revathy, M P Balasubramanian
ABSTRACT
The role of limonin as potent anti carcinogenic, apoptosis and chemotherapeutic agents has been supported by limited studies. In this study, limonin is identified as a potent anti proliferative agent against human hepatoma HepG2 cells based on the cell viability study, LDH leakage assay. Induction of apoptosis in HepG2 cells by limonin was evidenced by western blot analysis of Bax, Cyclin D1, Caspase 3 and Caspase9. Since Wnt signalling is involved in the initiation and sustaining of hepatocellular carcinoma we studied differential expression of LRP5, LRP6 and DKK wnt players. Limonin found to down regulate these players which forms a rationale for further investigation on effect on limonin in cancer therapy.
Sigma-Aldrich
Limonin, from citrus seeds, >90% (HPLC)