MilliporeSigma
  • A novel function for tissue inhibitor of metalloproteinases-3 (TIMP3): inhibition of angiogenesis by blockage of VEGF binding to VEGF receptor-2.
A novel function for tissue inhibitor of metalloproteinases-3 (TIMP3): inhibition of angiogenesis by blockage of VEGF binding to VEGF receptor-2.
Nature medicine (2003-03-26)
Jian Hua Qi, Quteba Ebrahem, Nina Moore, Gillian Murphy, Lena Claesson-Welsh, Mark Bond, Andrew Baker, Bela Anand-Apte
ABSTRACT
Tissue inhibitor of metalloproteinases-3 (TIMP3) is one of four members of a family of proteins that were originally classified according to their ability to inhibit matrix metalloproteinases (MMP). TIMP3, which encodes a potent angiogenesis inhibitor, is mutated in Sorsby fundus dystrophy, a macular degenerative disease with submacular choroidal neovascularization. In this study we demonstrate the ability of TIMP3 to inhibit vascular endothelial factor (VEGF)-mediated angiogenesis and identify the potential mechanism by which this occurs: TIMP3 blocks the binding of VEGF to VEGF receptor-2 and inhibits downstream signaling and angiogenesis. This property seems to be independent of its MMP-inhibitory activity, indicating a new function for this molecule.