Merck
A MedChem toolbox for cereblon-directed PROTACs.
MedChemComm (2019-07-16)
Christian Steinebach, Izidor Sosič, Stefanie Lindner, Aleša Bricelj, Franziska Kohl, Yuen Lam Dora Ng, Marius Monschke, Karl G Wagner, Jan Krönke, Michael Gütschow
ABSTRACT
A modular chemistry toolbox was developed for cereblon-directed PROTACs. A variety of linkers was attached to a CRBN ligand via the 4-amino position of pomalidomide. We used linkers of different constitution to modulate physicochemical properties. We equipped one terminus of the linker with a set of functional groups, e.g. protected amines, protected carboxylic acids, alkynes, chloroalkanes, and protected alcohols, all of which are considered to be attractive for PROTAC design. We also highlight different opportunities for the expansion of the medicinal chemists' PROTAC toolbox towards heterobifunctional molecules, e.g. with biotin, fluorescent, hydrophobic and peptide tags.
Sigma-Aldrich
CRBN-6-5-5-VHL, ≥98% (HPLC)