MilliporeSigma
  • Chronic nicotine treatment decreases LPS signaling through NF-κB and TLR-4 modulation in the hippocampus.
Chronic nicotine treatment decreases LPS signaling through NF-κB and TLR-4 modulation in the hippocampus.
Neuroscience letters (2016-12-17)
Cecília Cerqueira Café-Mendes, Humberto Miguel Garay-Malpartida, Marília Brinati Malta, Larrissa de Sá Lima, Cristóforo Scavone, Zulma S Ferreira, Regina P Markus, Tania Marcourakis
ABSTRACT
The hippocampus is a brain region that is rich in nicotinic acetylcholine receptors (nAChRs), especially the α7 subtype. The hippocampus is severely affected in disorders that have a neuroinflammatory component, such as Alzheimer's disease, Parkinson's disease, and schizophrenia. Previous studies demonstrated both in vivo and in vitro that nicotine inhibits immunological responses, including those that are triggered by the inflammatory agent lipopolysaccharide (LPS), the endotoxin of Gram-negative bacteria. The present study investigated whether chronically administered nicotine interferes with the nuclear binding of nuclear factor-κB (NF-κB) and the expression of LPS-induced inflammatory response genes. The results indicated that chronic nicotine administration (0.1mg/kg, s.c., 14days) inhibited the LPS-induced nuclear binding of NF-κB and mRNA expression levels of Tnf, Il1b, Nos2, and Tlr4. The presence of both the selective α7 nAChR antagonist methyllycaconitine (MLA; 5.0mg/kg i.p., 14days) and the nonselective nAChR antagonist mecamylamine (Meca; 1.0mg/kg, s.c., 14days) reversed the inhibitory effects of nicotine. These results suggest that the chronic activation of α7- and αxβy-containing nAChRs reduces acute inflammatory responses in the brain.
Sigma-Aldrich
Methyllycaconitine citrate salt, from Delphinium brownii seeds, ≥96% (HPLC)