MilliporeSigma
  • Catechin gallates are NADP+-competitive inhibitors of glucose-6-phosphate dehydrogenase and other enzymes that employ NADP+ as a coenzyme.
Catechin gallates are NADP+-competitive inhibitors of glucose-6-phosphate dehydrogenase and other enzymes that employ NADP+ as a coenzyme.
Bioorganic & medicinal chemistry (2008-03-04)
Eui Seok Shin, Jiyoung Park, Jae-Min Shin, Dooho Cho, Si Young Cho, Dong Wook Shin, Mira Ham, Jae Bum Kim, Tae Ryong Lee
ABSTRACT
Recent studies have shown that glucose-6-phosphate dehydrogenase (G6PD) is an effectual therapeutic target for metabolic disorders, including obesity and diabetes. In this study, we used in silico and conventional screening approaches to identify putative inhibitors of G6PD and found that gallated catechins (EGCG, GCG, ECG, CG), but not ungallated catechins (ECG, GC, EC, C), were NADP(+)-competitive inhibitors of G6PD and other enzymes that employ NADP(+) as a coenzyme, such as IDH and 6PGD.
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Gallocatechin gallate, from green tea, ≥98% (HPLC)
Sigma-Aldrich
(−)-Epigallocatechin, ≥95% (HPLC), from green tea
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
(−)-Epicatechin gallate, ≥98% (HPLC), from green tea
Sigma-Aldrich
(−)-Epicatechin, ≥90% (HPLC)
Sigma-Aldrich
(−)-Catechin, ≥97% (HPLC), from green tea
Sigma-Aldrich
(−)-Catechin gallate, ≥98% (HPLC), from green tea
Supelco
(−)-Epicatechin, analytical standard
Sigma-Aldrich
(−)-Gallocatechin, ≥98% (HPLC)
Sigma-Aldrich
(−)-Epicatechin, ≥98% (HPLC), from green tea
Supelco
(−)-Epigallocatechin, analytical standard