Merck
  • Can Dasatinib Ameliorate the Hepatic changes, Induced by Long Term Western Diet, in Mice?
Can Dasatinib Ameliorate the Hepatic changes, Induced by Long Term Western Diet, in Mice?
Annals of anatomy = Anatomischer Anzeiger : official organ of the Anatomische Gesellschaft (2020-11-05)
Hassan Reda Hassan Elsayed, Mohammad El-Nablaway, Basma H Othman, Asim Mohammed Abdalla, Eman Mohammad El Nashar, Mostafa Mohammed Abd-Elmonem, Randa El-Gamal
ABSTRACT
Non-alcoholic fatty liver disease (NAFLD) is a worldwide disease that progresses into steatohepatitis (NASH) that has no current effective treatment. This study aimed, for the first time, to investigate the effect of Dasatinib; a tyrosine kinase inhibitor showing anti-PDGFR activity with a macrophage modulating efficacy, on NASH. NASH was induced, in C57BL/6 mice by western diet (WD). Control groups received either DMSO or Dasatinib. After 12 weeks, WD-fed mice received DMSO, Dasatinib (4 mg/kg) or Dasatinib (8 mg/kg) once daily, for four weeks. Serum was examined for ALT and lipid profile. Immunohistochemical staining for SREBP1 (lipogenesis marker), iNOS, arginase-1, CD68, CD163 (macrophage polarization markers), TGF-β (fibrosis marker) and ASMA (a marker for activated hepatic stellate cell), hepatic mRNA expression for SREBP-1, iNOS, arginase-1, TGF-β and PDGFA genes; and western blotting for phosphorylated PDGFR α and β, SREBP1, iNOS, arginase-1, IL1α, COX2, TGF-β and ASMA were performed. Liver sections were stained also for H & E, Oil red O and Sirius red. Dasatinib could ameliorate the WD-induced disturbance of serum ALT, lipid profile and significantly reduced hepatic expression of PDGFA, phosphorylated PDGFR α and β, IL1α, COX2, SREBP-1, iNOS, CD68, TGF-β and ASMA but increased expression for arginase-1 and CD163 (M2 macrophage markers). Moreover, Dasatinib reduced the steatosis, inflammation, hepatocellular ballooning, hepatic fibrosis and the high NAFLD activity scoring induced by WD. Dasatinib can prevent the progression of WD-induced NASH by attenuating lipogenesis, and inducing M2 macrophage polarization with antifibrotic activity.
Sigma-Aldrich
Dasatinib, ≥98% (HPLC)