MilliporeSigma
  • FOXP3+ associated with the pro-inflammatory regulatory T and T helper 17 effector cells in asthma patients.
FOXP3+ associated with the pro-inflammatory regulatory T and T helper 17 effector cells in asthma patients.
Experimental and therapeutic medicine (2016-10-06)
Jian-Guo Zhang, Xiao-Juan Chen, Tao Liu, Shu-Juan Jiang
ABSTRACT
Asthma is a chronic bronchial inflammation that results to reversible incidence of airway obstruction and shortness of breath. Under normal circumstances, the lung immune system is maintained in a state of controlled inflammation, where balance exists between protective immunity mediated by effector cells and tolerance mediated by cells with regulatory function. Therefore, the inflammation observed in asthma patients may be caused by an imbalance between regulatory T (Treg) cells (CD4-positive with high expression of CD25 surface markers) and forkhead box P3 (FOXP3)-positive pro-inflammatory T helper 17 (Th17) cells. The aim of the present study was to evaluate whether reduced Treg cells and increased Th17 cells could be observed in the peripheral blood samples of asthma patients. As important markers of Treg cells, the expression levels of FOXP3 and interleukin (IL)-17a were analyzed via reverse trancription-quantitative polymerase chain reaction. The results indicated that the levels of cytokines that promote Th17 cells, including IL-6, IL-23 and TGF-β, were found to increase in the bronchoalveolar lavage fluid sample of asthma patients. However, the IL-10 level in the corresponding sample was much lower compared with that in control individuals. In conclusion, these results suggest that asthma associated with a reduced proportion of Treg and Th17 cells in the blood is characterized by the expression of pro-inflammatory cytokines that may be beneficial for the continuous generation of Th17 cells.
Sigma-Aldrich
Human IL-10 ELISA Kit (for Lysates), for cell and tissue lysate samples
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
Estimated to ship on March 24, 2025
$24.50